Learn More
Sigma Organic Chemistry Sorbic acid calcium sal | 1G | MFCD00043805
Supplier: Sigma Organic Chemistry CDS0006081G

Sorbic acid calcium sal , 1G
About this Item:
MDL #: MFCD00043805
Molecular Weight: 262.32
UNSPSC Code: 12352200
Chemical Formula: C12H14CaO4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.