Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitan Monostearate, Spectrum™ Chemical
SDP

Catalog No. 18613975 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-975 500 g Poly Pail
18-613-974 2.5 kg Poly Pail
18-613-973 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-613-975 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SPA60500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1338-41-6

Chemical Identifiers

CAS 1338-41-6

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 500 g
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Sorbitan Monostearate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.