accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH(R)O(R)OH(S)OH

CAS RN 1338-41-6

IUPAC Name: 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl octadecanoate
Synonyms: sorbitan stearate
Molecular Weight (g/mol): 430.63
Molecular Formula: C24H46O6
InChi Key: HVUMOYIDDBPOLL-IIZJTUPISA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
Physical Form 
  • (2)
  • (10)
Grade 
  • (3)
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (10)
  • (10)
  • (2)
  • (2)
  • (7)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (10)
  • (3)
  • (4)
  • (3)
  • (3)
  • (4)