Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitol, Powder, NF, 91-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606961 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-961 2.5 kg Poly Bottle
18-606-960 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-606-961 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO2192.5KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SO219, 50-70-4, C6H14O6

Spectrum™ Chemical Sorbitol, Powder, NF is used in the pharmaceutical industry as a sweetener or humectant. Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

CAS Max % 1
pH 3.5 to 7.0
Boiling Point 295°C (563°F)
Ignition Residue 0.001
Packaging Poly Bottle
Quantity 2.5 kg
Percent Purity 91 to 100.5%
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Sorbitol Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.