Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitol Solution, USP, 70%, Spectrum™

Click to view available options
Quantity:
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
50-70-4,7732-18-5
C6H14O6
182.17
FBPFZTCFMRRESA-UHFFFAOYNA-N
hexane-1,2,3,4,5,6-hexol
OCC(O)C(O)C(O)C(O)CO
50-70-4,7732-18-5
C6H14O6
182.17
FBPFZTCFMRRESA-UHFFFAOYNA-N
hexane-1,2,3,4,5,6-hexol
OCC(O)C(O)C(O)C(O)CO

SO220, 50-70-4, C6H14O6

Spectrum™ Sorbitol Solution, 70 Percent, USP is a hyperosmotic laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Drum
Quantity 200 L
Ignition Residue 0.001
Water 28.5 -31.5%
CAS Max % 0.7,0.3
Grade USP