Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitol Solution, USP, 70%, Spectrum™ Chemical
SDP

Catalog No. 18606954 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-954 20 L Poly Drum
18-606-955 200 L Drum
18-606-951 6 x 500 mL Amber Glass Bottle
18-606-953 4 x 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-954 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO22020LTPL
Only null left
Add to Cart
Add to Cart

SO220, 50-70-4, C6H14O6

Spectrum™ Chemical Sorbitol Solution, 70 Percent, USP is a hyperosmotic laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-70-4 , 7732-18-5
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

CAS Max % 0.7, 0.3
Ignition Residue 0.001
Packaging Poly Drum
Quantity 20 L
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.