Learn More
Thermo Scientific™ Sorvall BIOS 16 Adapters
Catalog No. 75003342
Change view
Click to view available options
Tube Type:
5/7 mL Blood Collection Tube
500 mL Conical Bottle
Blood Collection
Conical
Standard Microplate or Deepwell Plate
No. of Places:
1
11
26
42
5 Microplates or 1 DW Plate
52
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tube Type No. of Places
75-003-342 Blood Collection 42
75-003-341 5/7 mL Blood Collection Tube 52
75-003-343 Conical 26
75-003-344 Conical 11
75-003-345 Standard Microplate or Deepwell Plate 5 Microplates or 1 DW Plate
75-003-346 500 mL Conical Bottle 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 75-003-342 Supplier Thermo Scientific™ Supplier No. 75003342
Add to Cart
Add to Cart
Adapters (set of 2 adapters) for Thermo Scientific Sorvall BIOS 16 centrifuges
Adapters dedicated to Thermo Scientific Sorvall BIOS 16 centrifuges, dedicated to large volume and bioprocessing applications. These adapters fit into the 2000 mL buckets of the 6x2000 mL rotor (Part Number 75003962) and 2000 mL buckets of the 8x2000 mL rotor (Part number 75003982) equipped with 2000 mL buckets (Part number 75003964). They are available as set of 2 adapters.
Thermo Scientific Sorvall BIOS 16 centrifuges include features that not only help enhance reliability, but make them much simpler to use. Whether you require outstanding capacity or high performance for application flexibility, they offer features that will help maximize productivity—and when combined with our space-saving footprint, one of these options is sure to be a perfect fit in your bioprocessing facility. These adapters fit into the 2000 mL buckets of the 6x2000 mL rotor (Part Number 75003962) and 2000 mL buckets of the 8x2000 mL rotor (Part number 75003982) equipped with 2000 mL buckets (Part number 75003964). These adapters are available as set of 2 adapters.
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type Blood Collection
For Use With (Equipment) Blood Tubes
No. of Places 42
Tube Size 10 mL
No. per Pack 2