Learn More
Thermo Scientific™ Sorvall BIOS 16 Adapters
Description
Adapters dedicated to Thermo Scientific Sorvall BIOS 16 centrifuges, dedicated to large volume and bioprocessing applications. These adapters fit into the 2000 mL buckets of the 6x2000 mL rotor (Part Number 75003962) and 2000 mL buckets of the 8x2000 mL rotor (Part number 75003982) equipped with 2000 mL buckets (Part number 75003964). They are available as set of 2 adapters.
Specifications
Specifications
| Type | Adapter |
| Tube Type | Standard Microplate or Deepwell Plate |
| Capacity | 5 Microplates or 1 DW Plate |
| For Use With (Equipment) | Microplates |
| No. of Places | 5 Microplates or 1 DW Plate |
| No. per Pack | 2 |