Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ SOS2 Recombinant Protein Antigen
SDP

Catalog No. NBP233487PE Shop All R&D Systems Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP233487PE 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. NBP233487PE missing translation for 'mfr' Novus Biologicals™ missing translation for 'supplierNo' NBP233487PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human SOS2. The SOS2 Recombinant Protein Antigen is derived from E. coli. The SOS2 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

This is a blocking peptide for NBP2-33487. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

Specifications

Gene ID (Entrez) 6655
Species Human
Purification Method Chromatography
Purity >80%
Concentration 0.5mg/mL
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Symbol SOS2
Label Type Unlabeled
Molecular Weight (g/mol) 26kDa
Product Type SOS2
Quantity 0.1mL
Regulatory Status RUO
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-33487. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Immunogen HPTPLEREPCKISFSRIAETELESTVSAPTSPNTPSTPPVSASSDLSVFLDVDLNSSCGSNSIFAPVLLPHSKSFFSSCGSL
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.