Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOSSC Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989107
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9107 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9107 Supplier Bioss Supplier No. BS9491R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen SOSSC
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene INIP
Gene Accession No. Q9NRY2
Gene Alias C9orf80; HSPC043; HSPC291; hSSB-interacting protein 1; hSSBIP1; INIP; INTS3 and NABP interacting protein; INTS3- and NABP-interacting protein; minute INTS3/hSSB-associated element; MISE; Sensor of single-strand DNA complex subunit C; Sensor of ssDNA subunit C; Single-stranded DNA-binding protein-interacting protein 1; SOSS complex subunit C; SOSSC; SOSS-C; SSB-interacting protein 1; SSBIP1
Gene Symbols INIP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SOSSC/C9orf80.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58493
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.