Learn More
ACROBiosystems Recombinant Protein;Human SOST / Sclerostin Protein;HEK293;100UG;HST,SOST,VBCH,Sclerostin
Supplier:  ACROBiosystems HSTH5245100UG
Encompass_Preferred
Recombinant Protein;Human SOST / Sclerostin Protein;100UG;Human SOST, His Tag (HST-H5245) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Tyr 213 (Accession # AAI01087.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 22.5 kDa. The protein migrates as 26-40 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Catalog No. 50-165-7747
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.