Learn More
Apexbio Technology LLC Tizanidine HCl 64461-82-1 100mg
SDP
Supplier:  Apexbio Technology LLC B1368100
Encompass_Preferred
Tizanidine hydrochloride (CAS 64461-82-1) is a small molecule that functions as an agonist at 2-adrenergic receptors By activating these presynaptic receptors it suppresses the release of neurotransmitters from central noradrenergic neurons thereby modulating synaptic transmission in the central nervous system This mechanism makes Tizanidine HCl a valuable tool for investigating 2-adrenergic signaling pathways neurotransmitter regulation and CNS synaptic function in biomedical research settings
Catalog No. 50-000-92522
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.