Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Sotalol hydrochloride
GREENER_CHOICE

Catalog No. p-7112851
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-525-0 50 mg
09-521-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 09-525-0 Supplier Tocris Bioscience™ Supplier No. 0952/50
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

β antagonist

Sotalol hydrochloride is a relatively potent pure β adrenergic antagonist, unique in possessing additional class III antiarrhythmic activity.

Specifications

Chemical Name or Material N-[4-[1-Hydroxy-2-[(1-methylethyl)amino]ethyl]phenyl]methanesulfonamide hydrochloride
CAS 959-24-0
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 50mM in water
Target Non-selective Adrenergic beta Receptor Antagonists
Molecular Formula C12H20N2O3S.HCl
Formula Weight Observed MW: 308.82
Purity 99%

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.