Learn More
Cayman Chemical SotalolhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1613610 MG
Encompass_Preferred
A pan-HDAC inhibitor (Kis 0.6-31 nM for HDAC1-11) inhibits growth in a panel of 37 human or mouse lung cancer cell lines including SCLC NSCLC and mesothelioma cells (IC50s 4-175 5-310 and 5-470 nM respectively) induces tumor regression in BK-T and RG-1 SCLC mouse xenograft models at 20 mg/kg i.p.
Catalog No. 50-297-1581
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.