Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ SoundControl™ SH for Full Brim Hat

Catalog No. 19002969 Shop All MSA Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
25 dB
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Noise Reduction Rating (NRR)
19-002-969 25 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-002-969 Supplier MSA™ Supplier No. 10129327
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SoundControl™ SH Ear Muffs are designed to fit snugly into the accessory slots on the MSA V-Gard™ Slotted Hard Hat.

  • Spring design offers low pressure and a high level of comfort
  • Three distinct wearing positions for ease of use
  • Excellent grip makes it easy to adjust the cups, even when wearing gloves
  • Provides maximum space for the ears inside the cup
  • Soft ear cup material for extended wear
  • Easy ear cup height adjustment
  • Retain the Class E dielectric rating on the V-Gard Full Brim Hard Hat

Specifications

Color Gray
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Equipment) MSA™ Full Brim V-Gard™ Hard Hat
For Use With (Application) Confined Space, Working at Heights, Welding
Model 10129327
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 25 dB
Product Type SoundControl
Dielectric Yes
Style Hard Hat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.