Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ SoundControl™ HPE Ear Muffs

Catalog No. 19171020 Shop All MSA Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
26 dB
Style:
Over-the-head
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Noise Reduction Rating (NRR) Style
19-171-020 26 dB Over-the-head
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-171-020 Supplier MSA™ Supplier No. 10061271
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SoundControl™ HPE Ear Muffs provide high-rated protection for low-frequency environments while maintaing maximum comfort.

  • Provides high-rated protection for low-frequency environments
  • Designed specifically for very high noise levels
  • Padded headband on over-the-head style
  • Cap style to be used with MSA Slotted Hard Hats
  • Super-soft ear cushions provide all-day comfort
  • Easily adjustable headband force

Specifications

Color Black, Gray
ANSI S3.1974 Yes
CSA Z94.2 Class A
Cushion Color Black
For Use With (Application) Airfields, confined space, mining, sanding and grinding, food processes, welding, power plants
High Visibility No
Model 10061271
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 26 dB
Product Type SoundControl
Headband Color Black
Dielectric No
Style Over-the-head
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.