Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva SOURCE™ 15Q

Catalog No. 45002028 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-028 50 mL
45-002-027 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-028 Supplier Cytiva Supplier No. 17094701
Add to Cart
Add to Cart

Strong anion exchanger with high chemical stability for wide range of working conditions with good resistance to cleaning conditions at high pH

  • Bioprocess medium
  • Ligand: quaternary ammonium
  • Average particle size: 15μm
  • Matrix: polystyrene/divinylbenzene
  • pH stability working range: 2 to 12
  • For polishing steps in both research and industrial applications
  • Available in 10mL and 50mL bottles

TRUSTED_SUSTAINABILITY

Specifications

Buffer 20mM N-methylpiperazine, 20mM Piperazine, 20mM Bis-Tris, 20mM Triethanolamine, 20mM Tris, 20mM Diethanolamine, 20mM Ethanolamine, Glycine, 20mM 1,3-diamino-propane, 20mM Piperidine
Product Type Source 15Q chromato graphy media
Quantity 50 mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate 1800 cm/hr.
Ligand Type Quaternary Ammonium
Max. Pressure 5 bar
pH Range 2 to 12
Particle Size 15μm
Support Type Polystyrene/Divinylbenzene
Content And Storage 4° to 30°C
For Use With (Application) Polishing steps in both research and industrial applications
Matrix 15Q Chromatography Media
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.