Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva SOURCE™ 30Q Chromatography Media

Catalog No. 45002029 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-029 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-029 Supplier Cytiva Supplier No. 17127501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polymeric, strong anion exchanger designed for intermediate purification and large-scale polishing of proteins, peptides and oligonucleotides

  • Mono-sized 30μm rigid polystyrene/divinyl benzene polymer matrix, modified with quaternary ammonium (Q) strong anion exchange groups
  • Low back pressure and high flow rates for high productivity
  • High chemical stability for wide range of working conditions

TRUSTED_SUSTAINABILITY

Specifications

Buffer All common ion exchange Buffers
Product Type Source 30Q chromato graphy media
Quantity 50mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate 2000cm/hr.
Ligand Type Quaternary Ammonium
Max. Pressure 145psi
pH Range 2 to 12
Particle Size 30μm
Support Type Polystyrene/Divinylbenzene
Content And Storage 4° to 30°C
For Use With (Application) Purification and large-scale polishing
Matrix 15Q Chromatography Media
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.