Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ ApoScreen™ Annexin V Apoptosis Kit
SDP

Catalog No. OB1001002 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB10010-02 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB10010-02 Supplier SouthernBiotech™ Supplier No. 1001002
Add to Cart
Add to Cart

Includes

1mL annexin V-FITC, 1mL propidium iodide, and 7 x 1.5mL 10X binding buffer

Evaluate subpopulations of cells undergoing apoptosis, using fluoroscein-labeled annexin V and propidium iodide. Superior to DNA-based assays for the early detection of cells in the apoptic pathway.

  • Double staining procedure distinguishes live, necrotic and apoptotic cells
  • Labeled annexin V can also be used to analyze changes in membrane asymmetry in platelets and erythrocytes
  • Analysis can be performed on any flow cytometer equipped with a single laser using excitation at 488nm
  • Cells can be examined with a fluorescence microscope equipped with filters for discrimination of FITC and rhodamine/PI
  • Individual ApoScreen labeled annexin V reagents

Specifications

Quantity 1 Kit
Sample Volume 1 mL
Content And Storage 2°C to 8°C
For Use With (Application) Detection of cells in the apoptic pathway
Conjugate FITC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.