Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Aposcreen Annexin V-Biot
SDP

Catalog No. OB1003908 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB1003908 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB1003908 Supplier SouthernBiotech™ Supplier No. 1003908

missing translation for 'validMainframeMsg'

Flow Cytometry, Fluorescence microscopy

Specifications

Quantity 100 Tests
pH 7.4
Sample Volume 1 mL
Conjugate Biotin
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.