Learn More
Description
Includes
Enzyme-labeled, affinity-purified goat antibodies to mouse Ig (polyvalent) for initial screening; Antibodies to mouse IgM, IgG1, IgG2a, IgG2b, IgG3, kappa, and lambda for characterizing clones that test positive; Unlabeled goat anti-mouse Ig capture antibody; Substrate (Mfr. No. 5300-04: p-nitrophenyl phosphate; Mfr. No. 5300-05: 2,2-azino-di-[3-ethylbenzthiazoline sulfonic acid] [ABTS]); Substrates also available separately
- Ideal for screening and characterizing murine hybridomas
- Use either System with Mouse Immunoglobulin Isotype Panel (Mfr. No. 5300-01) for quantifying immunoglobulin levels in hybridoma supernatants and mouse serum
Specifications
Specifications
| Product Type | ELISA Kit |
| Quantity | 1 Kit, >1000 Tests |
| Species | Mouse |
| Conjugate | HRP |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | For system with Mouse immunoglobulin isotype panel |
| Content And Storage | 2°C to 8°C |
| Research Area | Antibody Isotyping |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
