Learn More
SouthernBiotech™ Genetically Distinct Collagens: Bovine Type I
Shop All Southern Biotechnology Associates Products

Description
Bovine origin, Type I.
- Can be used as a collagen standard, antigen for antibody production, and coating material in cell culture studies, and in the formation of collagen gels
- Prepared via controlled and limited pepsin digestion; greater than 90% pure
- Lyophilized
Specifications
Specifications
| For Use With (Application) | Cell Culture Coating Material, Antigen for Antibody Production, Collagen Gel Formation, Collagen Standard |
| Purification Method | Controlled and limited pepsin digestion, followed by selective salt precipitation |
| Quantity | 0.25 mg |
| Source | Bovine |
| Product Type | Collagen, Bovine Type I |
| Content And Storage | Store at 2°C to 8°C or in small aliquots at -20°C |
| Form | Lyophilized |
| Purity or Quality Grade | >90% by SDS-PAGE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.