Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Genetically Distinct Collagens: Bovine Type II
SDP

Catalog No. OB122002S Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.25 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB1220-02S 0.5 mg
OB1220-02 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OB1220-02S Supplier SouthernBiotech™ Supplier No. 122002S
Add to Cart
Add to Cart

Highly purified fibrous protein for a variety of applications

Bovine origin, Type II.

  • Can be used as a collagen standard, antigen for antibody production, and coating material in cell culture studies, and in the formation of collagen gels
  • Prepared via controlled and limited pepsin digestion; greater than 90% pure
  • Supplied as a 0.5mg/mL solution in 500mmol/L acetic acid or lyophilized

Specifications

Concentration 0.5 mg/mL
For Use With (Application) Collagen Gel Formation, Antigen for Antibody Production, Collagen Gel Formation, Collagen Standard
Purification Method Controlled and limited pepsin digestion, followed by selective salt precipitation
Quantity 0.5 mg
Source Bovine
Product Type Collagen, Bovine Type II
Content And Storage Store at 2°C to 8°C or in small aliquots at -20°C
Form Liquid
Purity or Quality Grade >90% by SDS-PAGE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.