Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Goat Fibronectin
SDP

Catalog No. OB143001 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB1430-01 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB1430-01 Supplier SouthernBiotech™ Supplier No. 143001
Add to Cart
Add to Cart

Purity >90% as judged by SDS-PAGE analysis (MW 50 kDa)

  • Fresh goat plasma
  • Purification by affinity chromatography on gelatin covalently linked to agarose

Specifications

For Use With (Application) Antigen for Antibody Production, Cell Culture Coating Material, Fibronectin Standard
Formulation Purified fibronectin may be reconstituted in 4M Urea solution, or dissolved in Cyclohexylaminopropanesulfonic Acid (CAPS) Buffer at pH 11
Molecular Weight (g/mol) 220
Name Goat Fibronectin
pH Range 11
Purification Method Affinity Chromatography
Quality Control Testing >90% by SDS-PAGE
Quantity 1 mg
Source Goat Plasma
Storage Requirements -20°C
Cross Reactivity Goat
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.