Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Mouse, Unconjugated, Polyclonal, Isotype Control, Southern Biotech™
SDP

Catalog No. OB010701 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB0107-01 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB0107-01 Supplier Southern Biotechnology Associates Supplier No. 010701
Add to Cart
Add to Cart

Mouse Polyclonal Isotype Control

SouthernBiotech isotype controls and purified immunoglobulins are useful as negative controls and standards in a variety of immunoassays.

Specifications

Applications Blocking Assay, ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, Western Blot
Classification Polyclonal
Concentration 5mg/mL
Conjugate Unconjugated
Formulation Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
Host Species Mouse
Quantity 10 mg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 2-8°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.