Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Mouse Laminin
SDP

Catalog No. OB141501 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB1415-01 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB1415-01 Supplier SouthernBiotech™ Supplier No. 141501
Add to Cart
Add to Cart

Purity >90% as judged by SDS-PAGE analysis

  • EHS sarcoma source
  • The extract is purified by salt precipitation, ion exchange and gel filtration chromatography

Specifications

Concentration 1mg/mL
For Use With (Application) Antigen for Antibody Production, Cell Culture Coating Material, Laminin Standard
Formulation Purified Protein is supplied as a 1mg/mL solution in 1mL of 0.4M NaCl, 0.05M Tris HCl, pH 7.4
Name Mouse Laminin
pH Range 7.4
Purification Method Gel Filtration/Ion Exchange Chromatography
Quality Control Testing >90% by SDS-PAGE
Quantity 1 mg
Source EHS Sarcoma
Storage Requirements 2° to 8°C
Cross Reactivity Mouse
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.