Learn More
Description
- Source: Placental villi
- Purification: Controlled and limited pepsin digestion, followed by selective salt precipitation
- Purity: >90% as judged by SDS-PAGE analysis
- Type I collagen standard
Antigen for antibody production; Coating material for cell culture studies; Formation of collagen gels
Specifications
Specifications
| For Use With (Application) | Antibody Production, Cell Culture (Coating Material), Collagen Gel Formation |
| Quantity | 0.5 mg |
| Purity | >90% |
| Concentration | 0.5 mg/mL |
| Purification Method | Controlled and limited pepsin digestion, followed by selective salt precipitation |
| Source | Bovine |
| Content And Storage | 2°C to 8°C |
| Product Type | Collagen |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
