Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Type III Bovine Collagen Protein
SDP

Catalog No. OB124002S Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB124002S 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB124002S Supplier SouthernBiotech™ Supplier No. 124002S
Add to Cart
Add to Cart

To insure lot to lot consistency, each batch of product is tested to conform to characteristics of a standard reference reagent using SDS-PAGE analysis

  • Source: Placental villi
  • Purification: Controlled and limited pepsin digestion, followed by selective salt precipitation.
  • Purity: >90% as judged by SDS-PAGE analysis

Type III collagen standard; Antigen for antibody production; Coating material for cell culture studies; Formation of collagen gels

Specifications

For Use With (Application) Antibody Production, Cell Culture (Coating Material), Collagen Gel Formation
Quantity 0.5 mg
Purity >90%
Concentration 0.5 mg/mL
Purification Method Controlled and limited pepsin digestion, followed by selective salt precipitation
Source Bovine
Content And Storage 2°C to 8°C
Product Type Collagen
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.