Learn More
Medchemexpress LLC Sovleplenib (Synonyms: HMPL-523) 100mg, Purity: 99.56%, CAS No. : 1415792-84-5
SDP
Supplier:  Medchemexpress LLC HY145598100MG
Encompass_Preferred
Sovleplenib (HMPL-523) is a highly potent, orally available and selective SYK inhibitor with an IC50 of 25 nM. Anti-tumor activity. Sovleplenib can be used for the research of immune thrombocytopenia (ITP).
Catalog No. NC2891618
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.