Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX-1 Polyclonal Antibody, Abbomax

Catalog No. 502203982
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3982 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3982 Supplier Abbomax Inc Supplier No. 601480
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Mouse embryonic brain Cellular location: Nuclear.

SOX-1 Polyclonal antibody specifically detects SOX-1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SOX-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene SOX1
Gene Accession No. O00570, P53783
Gene Alias BB176347; LOC100912302; sex determining region Y-box 1; Sox1; Sox-1; sox1.S; SRY (sex determining region Y)-box 1; SRY box 1; SRY-box 1; SRY-box 1 S homeolog; SRY-box containing gene 1; SRY-box containing protein; SRY-box transcription factor 1; SRY-related HMG-box gene 1; transcription factor SOX-1; transcription factor SOX-1-like; XELAEV_18015596mg; XlSox1; xSox1
Host Species Rabbit
Immunogen A synthetic peptide derived from N-terminus of Human SOX-1 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20664, 6656
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.