Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523351
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523351 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523351 Supplier Invitrogen™ Supplier No. PA523351
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SOX1 Polyclonal Antibody for Western Blot

SOX1, also known as SRY (sex determining region Y)-box 1, is a transcription factor belonging to the SRY-related HMG box family of transcription factors with a HMG box DNA-binding domain. SOX1 works through multiple independent pathways to promote neuronal cell fate determination and differentiation. This nuclear protein acts as a modulator of LINE retroposons promoter activity and is also involved in the development of the central nervous system, mainly neural and lens development. Transcriptional activator SOX1 binds directly to the HES1 promoter and suppresses HES1 transcription, thus attenuating Notch signaling. It also binds to beta-catenin and suppresses beta-catenin-mediated TCF/LEF signaling, thus potentially attenuating the WNT signaling pathway. Expression of SOX1 is seen mostly in embryonic tissues and moderately in brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene SOX1
Gene Accession No. O00570, P53783, Q2PG84
Gene Alias BB176347; LOC100912302; sex determining region Y-box 1; Sox1; Sox-1; sox1.S; SRY (sex determining region Y)-box 1; SRY box 1; SRY-box 1; SRY-box 1 S homeolog; SRY-box containing gene 1; SRY-box containing protein; SRY-box transcription factor 1; SRY-related HMG-box gene 1; transcription factor SOX-1; transcription factor SOX-1-like; XELAEV_18015596mg; XlSox1; xSox1
Gene Symbols SOX1, sox1.S
Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human SOX1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20664, 6656, 734174, 779569
Target Species Human, Mouse, Monkey, Horse, Frog, Marsupial
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.