Learn More
SOX12 Monoclonal Antibody (4A9)

Description
SOX12 Monoclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | SOX12 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4A9 |
| Concentration | 0.2 to 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SOX12 |
| Gene Accession No. | O15370 |
| Gene Alias | 2010205A02Rik; Protein SOX-22; SOX12; Sox-12; SOX22; SOX-22 protein; SRY (sex determining region Y)-box 12; SRY box 12; SRY-box 12; SRY-box containing gene 12; SRY-related HMG-box gene 22; transcription factor SOX-12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.