Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX17 Monoclonal Antibody (OTI3H5)
GREENER_CHOICE

Catalog No. PIMA524886
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA524886 100 μL
1 options

Catalog No. PIMA524886

Supplier: Thermo Scientific MA524886

Only null left
Add to Cart
Add to Cart

SOX17 Monoclonal Antibody (OTI3H5)

SOX17 Monoclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX17
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3H5
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SOX17
Gene Accession No. Q9H6I2
Gene Alias HMG transcription factor SOX17; HMG-box transcription factor Sox17; Sox17; Sox-17; SRY (sex determining region Y)-box 17; SRY box 17; SRY-box 17; SRY-box containing gene 17; SRY-box transcription factor 17; SRY-related HMG-box transcription factor SOX17; transcription factor SOX-17; transcription factor SOX-17; LOW QUALITY PROTEIN: transcription factor SOX-17; VUR3
Gene Symbols SOX17
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 177-414 of SOX17 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 486955, 64321
Target Species Human, Canine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.