Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX6 Monoclonal Antibody (OTI1E7), TrueMAB™, OriGene
SDP

Catalog No. 501682463
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2463 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2463 Supplier OriGene Supplier No. TA808919
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transcriptional activator. Binds specifically to the DNA sequence 5′-AACAAT-3′. Plays a key role in several developmental processes, including neurogenesis and skeleton formation.

Specifications

Antigen SOX6
Applications Western Blot
Classification Monoclonal
Clone OTI1E7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SOX6
Gene Accession No. P35712
Gene Alias HSSOX6, SOXD
Gene Symbols SOX6
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-244 of human SOX6 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55553
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.