Learn More
Invitrogen™ SOX9 Recombinant Rabbit Monoclonal Antibody (SN74-09)
Shop All Thermo Scientific Products

Description
SOX9 Recombinant Monoclonal Antibody for Flow, ICC/IF, IHC (P), IP, Western Blot
Specifications
Specifications
| Antigen | SOX9 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | SN74-09 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | SOX9 |
| Gene Accession No. | P48436, Q04887 |
| Gene Alias | 2010306G03Rik; AV220920; CMD1; CMPD1; mKIAA4243; SOX 9; Sox9; SOX-9; Sox9 transcription factor; SRA1; SRXX2; SRXY10; SRY (sex determining region Y)-box 9; SRY (sex determining region Y)-box 9 (campomelic dysplasia, autosomal sex-reversal); SRY (sex determining region Y)-box9; SRY (sex-determining region Y)-box 9 protein; SRY box 9; SRY-box 9; SRY-Box 9 ; SRY-box containing 9; SRY-box containing gene 9; SRY-box containing protein 9; SRY-box transcription factor 9; SRY-related HMG-box, gene 9; transcription activator Sox9; transcription factor SOX-9 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.