Learn More
3M™ Soy Protein Rapid Kit L25SOY, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S26920 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Soy Protein Rapid Kit L25SOY, Sold by AquaPhoenix Scientific
Catalog No.
S26920 Soy Protein Rapid Kit L25SOY, Sold by AquaPhoenix Scientific
1 options
Catalog No. S26920 Supplier 3M™ Supplier No. L25SOY
Add to Cart
Add to Cart
For screening for the presence of soy proteins in clean-in-place (CIP) final rinse water, environmental swab samples, food ingredients and processed food products
  • Results visible at 11 ± 1 min. after application of sample
  • Rapid, qualitative immunochromatographic lateral flow test cartridges designed for detection of allergenic food proteins
  • Designed to detect both processed and unprocessed allergen proteins.
  • Test can accommodate environmental swab, CIP rinse water and food samples
Specifications
Format Kit
Product Type Soy Protein Rapid Kit
Test Type Allergen
For Use With (Application) Screening soy proteins in CIP final rinse water, environmental swab samples, food ingredients and processed food products
Quantity 25/Cs.