Learn More
Selleck Chemical LLC SP 600125-E1391-5MG
SDP
Supplier:  Selleck Chemical LLC E13915MG
Encompass_Preferred
SP 600125 negative control (JNK inhibitor II negative control) is a methylated analog of SP 600125 and inhibits JNK2 and JNK3 with IC50 of 18 M and 24 M respectively
Catalog No. 50-313-6583