Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP Scientific Glassware Washers

Catalog No. 15365201
Change view
Click to view available options
Includes:
2 x Di rinse
One spindle-ready chamber
Two spindle-ready chambers
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
15-365-201 One spindle-ready chamber
15-365-200 2 x Di rinse
15-365-202 Two spindle-ready chambers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 15-365-201 Supplier SP Scientific Supplier No. NLW1395DS
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
  • Two independently operated chambers with up to four loading levels for high volume washing
  • Multiple cycle selections and microprocessor control systems ensure that each load is processed with minimum energy consumption
  • Delayed start feature allows washer to operate unattended during off-peak hours to reduce energy costs
  • Temperature selection to 70°C (158°F) for optimal cleaning of different loads
  • Spindle-ready models can accommodate direct injection spindle racks (sold separately) for narrow-neck glassware
  • Three separate filters protect pumps from debris and are conveniently located for easy access
  • 304 stainless steel chamber and exterior casing looks elegant and ensures long life with protection from corrosion
  • Equipped with noise buffering insulation for quieter run cycles
  • Efficient, minimal water consumption with spotless results
  • DI models easily install with fixed or movable laboratory DI water systems; optional DI Booster Pump recommended when the incoming DI water source has a pressure of less than 18psi
  • Enhanced dual fan drying system
  • Top and bottom stainless steel open racks accommodate a variety of glassware and accessories and are removable
  • Stainless steel baskets and other accessories hold tubes, bottles, petri dishes and more
  • 3 in. lockable casters allow easy, controlled unit mobility
  • MET Labs to UL 61010-1 & CSA 22.2 61010-1 certified
  • Features 6 Cycle Selections including a Customizable Timed Option. All Wash cycles have pre-wash, wash, pre-rinse and rinse cycles to prevent contamination.
    • Rinse Only - Short tap water rinse cycle
    • Normal Wash - Lower temperature (55°C) longer cycle with three (3) final rinse cycles
    • Daily Wash - Standard temperature (65°C) with two (2) final rinse cycles
    • Heavy Wash - Higher temperature (70°C) longer cycle with multiple pre-wash and final rinse cycles
    • Heavy Wash Time Saver - Lower temperature (55°C) version of Heavy Wash with a shorter overall cycle time
    • Timed Wash Programs - Standard temperature (65°C) in programmable 15 minute increments up to 2.5 hours

Specifications

Tiers Vertical
Certifications/Compliance ETL and ETL-C
Includes One spindle-ready chamber
Height (English) 78.5 in.
Height (Metric) 199.4 cm
Width (English) 26.5 in.
Weight (English) Shipping 370 lb.
Weight (Metric) Shipping 168 kg
Amperage 15 A
Voltage 2 x 115 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.