Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wilmad-LabGlass™ SP Scienceware™ 10mm OD High-Throughput NMR Tubes

Catalog No. 16800181 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Length (Metric):
17.8 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
16-800-181 17.8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Note: For NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, please refer to Wilmad Precision NMR Tubes.
Catalog No. 16-800-181 Supplier Wilmad-LabGlass™ Supplier No. 660000081
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Note: For NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, please refer to Wilmad Precision NMR Tubes.

Designed for routine use with small molecule (MW<500) samples, Wilmad-LabGlass 10mm OD High-Throughput NMR tubes are ideal for use in most low to mid field NMR spectrometers (up to 600 MHz).

  • Manufactured from borosilicate glass that meets ASTM Type 1 Class B standard and recognized as N51A
  • This glass may contain Fe2O3 which is paramagnetic and can impact shimming quality at high field
  • Guaranteed spectral resolution for small molecule (MW<500) samples up to 600 MHz
  • Averaged camber of 60 micron
  • Ideal for MRI imaging applications
  • Blue disposable NMR caps included; other colors available separately
  • 100% inspected for surface defects and physical dimension to ensure the success of your experiments

Specifications

Camber (Metric) 60 μm
Product Type High-Throughput NMR Tube
Material Borosilicate Glass
For Use With (Equipment) Low to mid field NMR spectrometers (up to 600 MHz)
Length (Metric) 17.8 cm
Color Clear
Diameter (Metric) Outer 9.944±0.025 mm
For Use With (Application) Small molecule (MW<500) samples
Includes Blue disposable NMR caps
Length (English) 7 in.
Thickness (English) Wall 0.02 in.
Thickness (Metric) Wall 0.60 mm
Quantity 100
Model WG-4000-7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.