Learn More
Bel-Art™ SP Scienceware™ Accredited Hydrometer Calibration Services: ISO 17025:2005
Catalog No. 13202708 Shop All H-B Instrument Company Products
Change view
Click to view available options
Calibration Points:
1 Measuring Point
2 Measuring Points
3 Measuring Points
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Points
13202708 2 Measuring Points
13-202-705 1 Measuring Point
13202709 3 Measuring Points
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13202708 Supplier Bel-Art™ Supplier No. B601110000
Add to Cart
Add to Cart
Meets the requirements of ISO, ANSI and NIST standards. Bel-Art™ SP Scienceware™ Accredited Hydrometer Calibration Services; ISO 17025:2005 can be used to calibrate new hydrometers as well as to recalibrate previously calibrated instruments. Report includes correction values.
Properly calibrated instruments are required to adhere to laboratory standard operating procedures. Many factors can cause drift from accepted values. In general laboratory use, it is recommended that hydrometers be calibrated at one year intervals.
  • Accredited calibration documentation meets the requirements of ISO 17025 and ANSI NCSL-Z-540
  • Calibration is performed in SP Scienceware's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01)
  • Calibration is performed against equipment whose calibration is traceable to NIST
  • Average 10 day turn-around from receipt of the instrument for hydrometer calibration
  • The calibration report with correction values will be sent and will also be stored in an SP Scienceware database
  • A 1-year recalibration reminder will be sent to ensure reliable and accurate instrument readings throughout the life of the equipment
Instructions for Ordering/Receiving Calibration Services:
  1. Contact Fisher Scientific at 800-766-7000 to let them know the type of instrument you will be sending in, or the catalog number of the item that you would like to order, and the required calibration points. The representative will provide you with the cost and place the order.
  2. If sending in a piece of equipment, the representative will provide you with a Return Material Authorization (RMA) number. Securely pack and send your instrument(s) with the RMA number on the outside of the box to: H-B Instrument – SP Scienceware, 102 W Seventh Ave, Trappe, PA 19426, Attn. Calibration.
  3. SP Scienceware™ will perform the calibration and send you the calibration report along with the instrument.
Specifications
For Use With (Equipment) Hydrometers
Certifications/Compliance ISO 17025, ANSI NCSL-Z-540, NIST
Description Accredited calibration/recalibration service for hydrometers
Calibration Points 2 Measuring Points
Duration 10 Day Turnaround (avg.)
Calibration Type Hydrometer
Includes Calibration report, data storage, 1 yr reminder
WARNING: Cancer - www.P65Warnings.ca.gov