Learn More
Bel-Art™ Biohazard Disposal Bags with or without Warning Label
Shop All SP Bel-Art & SP Wilmad-LabGlass Products

Description
- Transparent, polypropylene plastic biohazard bags are 0.03 mm (1.5 mil) thick
- High temperature autoclavable at a maximum of 135°C (275°F)
- May also be used as conventional trash can liners
Specifications
Specifications
| Certifications/Compliance | ASTM D1709-98 |
| Design | Biohazard Symbol w/ Sterilization Patch |
| Capacity (English) | 2.75 gal. |
| Color | Clear |
| Dimensions (L x W) | 12 x 24 in. |
| Disposable | Single-use |
| Length (Metric) | 30 cm |
| Width (Metric) | 61 cm |
| Autoclavable | Autoclavable |
| For Use With (Application) | Disposal of used pipettes, petri dishes, culture plates, membrane filters and more |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.