Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Biohazard Disposal Bags with or without Warning Label
SCI_ED

Catalog No. S43268 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Color:
Clear
Translucent
Design:
Biohazard Symbol w/ Sterilization Patch
Non-printed without Sterilization Patch
Dimensions (L x W):
12 x 24 in.
24 x 12 in.
24 x 30 in.
24 x 36 in.
30 x 24 in.
36 x 24 in.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Design Dimensions (L x W)
S43268 Clear Biohazard Symbol w/ Sterilization Patch 24 x 36 in.
S43261 Translucent Non-printed without Sterilization Patch 24 x 12 in.
S43263 Translucent Non-printed without Sterilization Patch 30 x 24 in.
S43258 Clear Biohazard Symbol w/ Sterilization Patch 12 x 24 in.
S43259 Clear Biohazard Symbol w/ Sterilization Patch 24 x 30 in.
S43264 Translucent Non-printed without Sterilization Patch 36 x 24 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. S43268 Supplier Bel-Art™ Supplier No. F131620009/EMD
This item is not returnable. View return policy

Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters,and other biohazard wastes

  • Transparent, polypropylene plastic biohazard bags are 0.03 mm (1.5 mil) thick
  • High temperature autoclavable at a maximum of 135°C (275°F)
  • May also be used as conventional trash can liners

Specifications

Certifications/Compliance ASTM D1709-98
Design Biohazard Symbol w/ Sterilization Patch
Capacity (English) 10.5 gal.
Color Clear
Dimensions (L x W) 24 x 36 in.
Disposable Single-use
Length (Metric) 61 cm
Width (Metric) 91 cm
Autoclavable Autoclavable
For Use With (Application) Disposal of used pipettes, petri dishes, culture plates, membrane filters and more
Width (English) 36 in.
Length (English) 24 in.
Thickness 1.5 mm
Material Polypropylene
Product Type Biohazard Waste Bag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.