Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel Art™ Polypropylene Sterilizing Trays and Covers
SCI_ED

Catalog No. S43344 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Capacity (Metric):
5 L
13 L
18 L
Type:
Sterilizing Tray
Tray Cover
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Type
S43344 5 L Tray Cover
S43341 5 L Sterilizing Tray
S43343 13 L Sterilizing Tray
S43348 18 L Sterilizing Tray
S43086 13 L Tray Cover
S43346 18 L Tray Cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S43344 Supplier Bel Art™ Supplier No. H162590000/EMD
Add to Cart
Add to Cart

Corrosion-proof polypropylene plastic tray and cover. Sterilizing trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.

  • Ideal for sterilizing instruments, bottles containing agar, deionized water, saline and other reagents, and for safely cleaning glassware in sinks
  • Polypropylene covers are 3.2mm (1/8 in.) thick with a 25.4mm (1 in.) skirt that fits snugly over the rim of the tray
  • Trays and lids are steam autoclavable at 121°C (250°F)

Specifications

Capacity (Metric) 5 L
Dimensions (L x W x H) 12 x 8 x 1 in. (30.5 x 20.3 x 2.54 cm)
Color White
Height (Metric) 2.54 cm
Length (Metric) 30.5 cm
Material Polypropylene
Type Tray Cover
Width (Metric) 20.3 cm
For Use With 5L Tray, Mfr. No. H162600000
Length (English) 12 in.
Height (English) 1 in.
Shape Rectangular
Thickness 3.2 mm
Width (English) 8 in.
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.