Learn More
Bel-Art™ Right-to-Know, Safety-Vented Wash Bottles with GHS Labeling
Shop All SP Bel-Art & SP Wilmad-LabGlass Products

Description
- Wide mouth 53 mm (2 in.) for easy refilling.
- Translucent LDPE bottles with color-coded caps for easy identification.
- Pre-printed with chemical name and formula, GHS pictogram and signal word, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.
- Bottles with labels that do not indicate a hazardous chemical do not have or require GHS pictogram or signal word.
- Methyl Ethyl Ketone and Toluene do not include vented valve.
- Assortment is 4 per bag, includes one each: Acetone, Isopropanol, Bleach, and Ethanol.
- Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm.
- Bottles with labels that do not indicate a hazardous chemical do not have or require GHS pictogram or signal word.
Compliance
Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA's HazCom 2012 requirements–as pertains to Workplace Labels
Specifications
Specifications
| Capacity | 16 oz., 500 mL |
| Capacity (Metric) | 500 mL |
| Capacity (English) | 16 oz. |
| Color | Yellow |
| Material | LDPE |
| Quantity | 4/Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Disposable | Reusable |
| Graduated | No |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.