Learn More
Bel Art™ Space Saver Vacuum Desiccator
SCI_ED
Catalog No. S43288 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Material:
Polypropylene, Polycarbonate
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S43288 -
S43283 Polypropylene, Polycarbonate
S43285 Polypropylene, Polycarbonate
S43284 -
S43286 -
S43287 Polypropylene, Polycarbonate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. S43288 Supplier Bel Art™ Supplier No. F420270000/EMD
Add to Cart
Add to Cart
This item is not returnable. View return policy
Degas liquids quickly and easily in the laboratory or classroom with these small plastic vacuum desiccators.
  • Strong, shatterproof units provide dry storage and protection for sensitive products
  • Lighter and less costly than similarly sized glass models; do not chip like glass
  • Hold a full vacuum (29.9 in. Hg) at room temperature for 24 hours
  • Tapered polypropylene stopcock with PTFE plug accepts tubing with 0.25 in. to 0.375 in. I.D. and allows a controlled entrance of air, preventing unnecessary turbulence
  • Neoprene O-ring assures a tight seal down to 0°C without the need for grease (ring will not be harmed if grease is used)
  • Flat Dome clear polycarbonate top with large ribbed knob maximizes interior clearances and provides an average of 13% greater interior volume over standard hemispherical domes
  • All models include a 0.125 in. (0.32cm) thick perforated plate; disposable or reusable desiccant can be placed on or under this plate
  • Clear polycarbonate bottom models offer increased viewing visibility from more angles
  • Models with white polypropylene bottoms can support incandescent crucibles on Minerit HD High Heat Ceramic Desiccator Plates (available separately, F42038 series)
  • Models with clear polycarbonate bottoms offer increased visibility from more angles
Certifications
Upper and lower sections, neoprene O-ring, stopcock, polypropylene sample plate
Specifications
Product Type Vacuum Desiccator
Diameter (Metric) Outer 27.3 cm