Learn More
Bel-Art™ Spinpak Octagonal Magnetic Stirring Bar Assortment, Restrainer
Shop All SP Bel-Art & SP Wilmad-LabGlass ProductsDescription
- Eight-sided Spinbar™ Octagonal Magnetic Stirring Bars provide greater surface area and added turbulence when compared to smooth cylindrical bars
- Integral pivot ring on each stirring bar aids in reducing friction and chattering
- Alnico magnet encapsulated in USP Class VI Teflon™ PTFE low friction coating; durable and inert; manufactured in registered ISO 9001:2008 facilities, as verified by SGS certification
- Included Spinbar Restrainer fits comfortably in one hand and retains Spinbar Magnetic Stirring Bars up to 80 mm (3 in.) long by holding them against vessel walls; eliminates splashing hazards when pouring
- Includes a variety of Spin bar sizes
Specifications
Specifications
| Type | Magnetic Stir Bar Set |
| Coating | PTFE, FDA-grade, USP Class VI |
| Autoclavable | Autoclavable |
| Shape | Octagon |
| Quantity | 11/Pk. |
| Includes | Two 0.31 O.D. x 0.50 in. L bars (white, red), two 0.31 O.D. x 1 in. L bars (white, yellow), two 0.31 O.D. x 1.5 in. L bars (white, blue), two 0.31 O.D. x 2 in. L bars (white, yellow), two 0.13 O.D. x 0.50 in. L bars (white, red), and one Restrainer |
| Material | Alnico Magnet |
| Certifications/Compliance | ISO 9001:2008 |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.