Learn More
Bel-Art™ Two-Color Wash Bottles, Safety-Vented™ and Safety Labeled, Wide-Mouth
Shop All SP Bel-Art & SP Wilmad Products

Description
A unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know Safety-Labeled Wash Bottles
- Pre-printed with chemical name and formula, the National Fire Protection Association (NFPA) section 704 four-color diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.
- Valve eliminates drips, allows dispensing of wash bottle in an upright or inverted position.
- Wide mouth 53 mm (2 in.) bottle for easy refilling.
- Polypropylene closure with leakproof design, color-coded to aid in easy identification.
- Translucent, LDPE solvent-resistant Sodium Hypochlorite is white LDPE for light-protection between 230 and 450.
- Toluene is red LDPE bottle.
- Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
- Labeled with six of the most commonly used solvent names or custom labeling available.
Order Info
8 oz. (250 mL)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Capacity | 16 oz., 500 mL |
| Capacity (Metric) | 500 mL |
| Color | Translucent |
| Diameter (Metric) Outer Neck | 53 mm |
| For Use With (Application) | Ethanol |
| Includes | Natural Polypropylene Cap |
| Material | Polyethylene |
| Quantity | 1 Ea. |
| Sterility | Non-sterile |
| Closure Color | Natural |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.