Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Two-Color Wash Bottles, Safety-Vented™ and Safety Labeled, Wide-Mouth
GSA_VA

Catalog No. 1139153 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
Chemical Label:
Acetone
Custom Write-on
Ethanol
Isopropanol
Methanol
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Chemical Label
11-391-53 250 mL Acetone
11-391-55 250 mL Isopropanol
11-391-56 250 mL Custom Write-on
01-189-0105 500 mL Ethanol
01-189-0106 500 mL Custom Write-on
01-189-0104 500 mL Isopropanol
11-391-54 250 mL Ethanol
11-391-57 250 mL Methanol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Use only with solvents compatible with LDPE.
Catalog No. 11-391-53 Supplier Bel-Art™ Supplier No. F116430222
Add to Cart
Add to Cart
Use only with solvents compatible with LDPE.

Unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know safety-labeled wash bottles.

A unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know Safety-Labeled Wash Bottles

  • Pre-printed with chemical name and formula, the National Fire Protection Association (NFPA) section 704 four-color diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.
  • Valve eliminates drips, allows dispensing of wash bottle in an upright or inverted position.
  • Wide mouth 53 mm (2 in.) bottle for easy refilling.
  • Polypropylene closure with leakproof design, color-coded to aid in easy identification.
  • Translucent, LDPE solvent-resistant Sodium Hypochlorite is white LDPE for light-protection between 230 and 450.
  • Toluene is red LDPE bottle.
  • Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
  • Labeled with six of the most commonly used solvent names or custom labeling available.

Order Info

8 oz. (250 mL)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Capacity 8 oz., 250 mL
Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Red
For Use With (Application) Acetone
Includes Wash Bottle
Material LDPE
Sterility Non-sterile
Closure Color Red
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Wide
Shape Cylindrical
Product Type Safety-vented Wash Bottle
Chemical Label Acetone
Closure Material Polypropylene
Autoclavable Not Autoclavable
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.