Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Wide Mouth Color-Code Safety Labeled Wash Bottles
GREENER_CHOICE

Catalog No. 22288650 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
For Use With (Application):
Acetone
Dichloromethane
Ethanol
Ethyl Acetate
Methyl Ethyl Ketone
Saline
Sodium Hypochlorite
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) For Use With (Application)
22-288650 1000 mL Acetone
22-273735 500 mL Dichloromethane
22-288641 500 mL Ethanol
22-273739 500 mL Methyl Ethyl Ketone
22-288639 500 mL Saline
22-293494 500 mL Ethyl Acetate
14-512-38 500 mL Sodium Hypochlorite
22-288654 1000 mL Ethanol
22-288648 1000 mL Methyl Ethyl Ketone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 22-288650 Supplier Bel-Art™ Supplier No. F116462232
Add to Cart
Add to Cart

Includes

Labeling with Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information

Easy-to-read labeling printed bottle promoting safe working environment

  • Wide mouth 53 mm (2 in.) bottle for easy refilling.
  • Made from solvent-resistant low-density polyethylene.
  • Polypropylene closure with leakproof design.
  • Pictogram instructions promote universal safety regardless of setting.
  • Use LYOB only with solvents compatible with LDPE.

Compliance

Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

Specifications

Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Clear
For Use With (Application) Acetone
Material LDPE
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Wide
Shape Cylindrical
Product Type Safety-labeled Wash Bottle
Chemical Label Acetone
Closure Material Polypropylene
Closure Size 53 mm
Autoclavable Not Autoclavable
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.