Learn More
Bel-Art™ Wide Mouth Color-Code Safety Labeled Wash Bottles
Shop All SP Bel-Art & SP Wilmad-LabGlass Products

Description
Includes
Labeling with Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information
Easy-to-read labeling printed bottle promoting safe working environment
- Wide mouth 53 mm (2 in.) bottle for easy refilling.
- Made from solvent-resistant low-density polyethylene.
- Polypropylene closure with leakproof design.
- Pictogram instructions promote universal safety regardless of setting.
- Use LYOB only with solvents compatible with LDPE.
Compliance
Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Specifications
Specifications
| Capacity | 16 oz., 500 mL |
| Capacity (Metric) | 500 mL |
| Capacity (English) | 16 oz. |
| Color | Clear |
| For Use With (Application) | Ethyl Acetate |
| Material | LDPE |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Disposable | Reusable |
| Graduated | No |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.